Learn More
Perkin Elmer US LLC Cassette for Avio 500 Torch Body Assembly
Catalog No. 502105534
Encompass_Preferred
This cassette is included in the Avio 500 Adjustable Torch Body Assembly (N0810128). This assembly also includes a Torch Mount (N0791285).
Catalog No. 50-210-5534 Supplier Perkin Elmer US LLC Supplier No. N0810214
This cassette is included in the Avio 500 Adjustable Torch Body Assembly (N0810128). This assembly also includes a Torch Mount (N0791285).