Learn More
Epredia™ Cassette II Biopsy Tissue Cassettes
Catalog No. B851738TN Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
B851738TN Tan
B851738BE Beige
B851738BL Blue
B851738GN Green
B851738GY Gray
B851738LA Lavender
B851738LG Light Green
B851738LI Lilac
B851738OR Orange
B851738PK Pink
B851738WH White
B851738YW Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. B851738TN Supplier Epredia™ Supplier No. B851738TN
Add to Cart
Add to Cart
For small tissue samples, these cassettes have a front-edge lid hinge with loose-packed lids that are available in 12 different color varieties
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
Specifications
Pore Style Small Squares
Packaging Type Loose Packed (Lids)
Quantity 1000/Cs.
Closure Included Yes
Closure Type Separate Lid, Front Hinge
PrintMate Compatible No
Color Tan
Product Type Cassette II with Color-Matched Lids