Learn More
Perkin Elmer US LLC Cassette Ion Lens Upgrade Kit for ELAN 6x00/9000/DRCs (Pre-2005)
Catalog No. 502113109
Encompass_Preferred
Demountable assembly that includes all mounting hardware, ShadowStop, and a Series II lens.
Catalog No. 50-211-3109 Supplier Perkin Elmer US LLC Supplier No. W1017871
Add to Cart
Add to Cart
Request a Quote
Demountable assembly that includes all mounting hardware, ShadowStop, and a Series II lens.