Learn More
Perkin Elmer US LLC Cassette Lens ShadowStop for ELAN 9000/DRCs (Post-2005)
Catalog No. 502104374
Encompass_Preferred
The PerkinElmer SCIEX patented ShadowStop prevents photons and un-ionized matrix constituents from entering the quadrupole, lowering system background, and eliminating drift. This innovation also enhances ease-of-use because the ShadowStop is grounded, making is a passive part of the ion optic design.
Catalog No. 50-210-4374 Supplier Perkin Elmer US LLC Supplier No. W1013361
The PerkinElmer SCIEX patented ShadowStop prevents photons and un-ionized matrix constituents from entering the quadrupole, lowering system background, and eliminating drift. This innovation also enhances ease-of-use because the ShadowStop is grounded, making is a passive part of the ion optic design.