Learn More
Mitegen Cassette Loading Station Insert for AutoGrid Cassette Puck Use
SDP
Catalog No. 5000494986
Encompass_Preferred
The MiTeGen Cassette Loading Station Insert is designed for easy AutoGrid Cassette Puck use in the ThermoFisher Scientific Cassette Loading station. It helps to facilitate the use of the pucks with the loading station.
Catalog No. 50-004-94986 Supplier Mitegen Supplier No. MCEMAGCPLSI1
The MiTeGen Cassette Loading Station Insert is designed for easy AutoGrid Cassette Puck use in the ThermoFisher Scientific Cassette Loading station. It helps to facilitate the use of the pucks with the loading station.