Learn More
Newcomer Supply Macrosette Tissue Cassettes, 250/box. Specially designed for larger tissue specimens (10mm deep).
SDP
Catalog No. NC0072902
Encompass
The Macrosette Tissue Processing & Embedding Cassettes are disposable plastic cassettes specially designed to hold larger tissue specimens during the tissue processing and embedding procedure, as well as easily fit into a standard cassette storage cabinet. The hinged lid comes attached and can be opened and closed as often as necessary, relocking securely without danger of specimen loss. There are large labeling areas on three sides of the cassette. Available in White. Dimensions: 1 9/16" x 1 1/8" x 1/2" H.
Catalog No. NC0072902 Supplier Newcomer Supply Supplier No. 5118W
The Macrosette Tissue Processing & Embedding Cassettes are disposable plastic cassettes specially designed to hold larger tissue specimens during the tissue processing and embedding procedure, as well as easily fit into a standard cassette storage cabinet. The hinged lid comes attached and can be opened and closed as often as necessary, relocking securely without danger of specimen loss. There are large labeling areas on three sides of the cassette. Available in White. Dimensions: 1 9/16" x 1 1/8" x 1/2" H.