Learn More
Electron Microscopy Sciences Air Sampling Cassette Opener
SDP
Catalog No. 5033885
Encompass
Heavy duty stainless steel cassette opener. Works with 25 mm, 37 mm and Air-O-Cell cassettes. No need to remove labels, this opener cuts right through and effortlessly pries cassettes open.
Catalog No. 50-338-85 Supplier Electron Microscopy Sciences Supplier No. 6523101
Heavy duty stainless steel cassette opener. Works with 25 mm, 37 mm and Air-O-Cell cassettes. No need to remove labels, this opener cuts right through and effortlessly pries cassettes open.