Learn More
Med Vet International Quick Shot X-ray Cassette, J0493X
Catalog No. 5000495288
Encompass_Preferred
This handy device holds the cassette at arm’s length directly attached to the portable x-ray unit. The "Quick Shot" allows a single person to take films and eliminates the need for assistance.Lightweight aluminum weighs only 2 1/2 lbs.Adjustable handle allows usage with all commercial portable x-ray units.Holds wide range of cassette and grid sizesNo guess work in achieving proper focal lengths"Bubble" level mounted on top of the handle and side of the cassette holderAdjustment knobs allow positioning of cassettes at all anglesFolds flat for easy storageFoam padded handle has holder for x-ray machine triggerRemoves many positioning variables
Catalog No. 50-004-95288 Supplier Med Vet International Supplier No. JOR493X
This handy device holds the cassette at arm’s length directly attached to the portable x-ray unit. The "Quick Shot" allows a single person to take films and eliminates the need for assistance.Lightweight aluminum weighs only 2 1/2 lbs.Adjustable handle allows usage with all commercial portable x-ray units.Holds wide range of cassette and grid sizesNo guess work in achieving proper focal lengths"Bubble" level mounted on top of the handle and side of the cassette holderAdjustment knobs allow positioning of cassettes at all anglesFolds flat for easy storageFoam padded handle has holder for x-ray machine triggerRemoves many positioning variables