Learn More
Electron Microscopy Sciences Air Sampling Cassette Sampling Stand, ABS-1
SDP
Catalog No. 5033887
Encompass
Sampling stand used to support sampling cassettes at elevated levels. ABS-1 Deluxe Sampling Stand: very similar to the economy stand but made in the USA from aluminum. Better durability and stability than the economy one.
Catalog No. 50-338-87 Supplier Electron Microscopy Sciences Supplier No. 65232ABS
Sampling stand used to support sampling cassettes at elevated levels. ABS-1 Deluxe Sampling Stand: very similar to the economy stand but made in the USA from aluminum. Better durability and stability than the economy one.