Learn More
Electron Microscopy Sciences Air Sampling Cassette Sampling Stand, Economy
SDP
Catalog No. 5033886
Encompass
Sampling stand used to support sampling cassettes at elevated levels. Sampling stand used to support sampling cassettes at elevated levels. ZA0042 Economy Sampling Stand: very nice stand, made from black anodized aluminum.
Catalog No. 50-338-86 Supplier Electron Microscopy Sciences Supplier No. 6523242
Sampling stand used to support sampling cassettes at elevated levels. Sampling stand used to support sampling cassettes at elevated levels. ZA0042 Economy Sampling Stand: very nice stand, made from black anodized aluminum.