Learn More
Electron Microscopy Sciences Slide/Cassette Storage File Box, 10/cs
SDP
Supplier:  Electron Microscopy Sciences 6328020
Encompass_Preferred
Moisture resistant, plastic covered corrugated boxes interlock to stack ten high, each supporting up to 500 lbs of overhead cassette and slide weight. Each box accommodates 2000 slides or 500 tissue blocks for varied storage requirements. Come unassembled, boxes require minimal storage space and can be easily assembled as needed.
Slide storage file with four compartments, each has one drawer, each drawer has one center divider, each division is 118" wide, accommodates up to 250 slides or 500 slides per drawer, or 2000 slides per file, 10 stackable files per case or 20,000 slides.
When these boxes are used as a cassette storage file, each compartment has two drawers, one on top of the other. Four compartments or eight drawers, each drawer accommodates 63 cassettes or 500 cassettes per file. 10 stackable files per case or 1500 cassettes. Cassette Storage File has 8 compartments, accommodates up to 500 tissue blocks.
Catalog No. 50-287-14