Learn More
Heidolph™ Cassette with Suspension for Hei-VAP Industrial Rotary Evaporator
SDP
Catalog No. 13900967 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Hei-VAP Industrial Rotary Evaporator
Catalog No. For Use With (Equipment)
13-900-967 Hei-VAP Industrial Rotary Evaporator
1 options
Catalog No. 13-900-967 Supplier Heidolph™ Supplier No. 036306101
Add to Cart
Add to Cart
Cassette with suspension guards against glassware breakage while providing a support for receiving flask. Includes attachment to HVI Rotavap
Included with Hei-VAP Industrial Rotary Evaporator
Specifications
Type Enclosure
For Use With (Equipment) Hei-VAP Industrial Rotary Evaporator
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov