Learn More
Thomas Scientific Cassette, Tissue Embedding with Attached Lid, 30ø Writing Area, AQUA, 500/Dispenser Box, 2 Boxes/Unit
SDP
Catalog No. 501460334
Encompass_Preferred
The 30Ý angled cassette features an extra-large writing surface and two side panels ideal for labeling. The cassette has an integral lid that snaps apart and securely locks into the base of the cassette. The lid can be removed and reattached repeatedly to access the specimen. Note: These cassettes are not suitable for use on cassette printers. Extra large textured writing surface Wide finger tab on the back of the cassette for easy opening The lid is designed to be removed from the base and reinserted into front tabs to lock into place
This item is not returnable. View return policy
Catalog No. 50-146-0334 Supplier Thomas Scientific Supplier No. 1202D72
The 30Ý angled cassette features an extra-large writing surface and two side panels ideal for labeling. The cassette has an integral lid that snaps apart and securely locks into the base of the cassette. The lid can be removed and reattached repeatedly to access the specimen. Note: These cassettes are not suitable for use on cassette printers. Extra large textured writing surface Wide finger tab on the back of the cassette for easy opening The lid is designed to be removed from the base and reinserted into front tabs to lock into place