Learn More
Thomas Scientific Cassette, Tissue Embedding with Attached Lid, 35ø Writing Area, BLUE, 500/Dispenser Box, 2 Boxes/Unit
SDP
Catalog No. 501460344
Encompass
The 35Ý angled cassette features a large writing surface and two side panels ideal for labeling. The one-piece design allows for the lids to be opened and closed repeatedly without the lid getting detached from the base. The lid securely locks into the base and keeps samples safe and secure. The 35Ý angled cassettes can be manually loaded into most cassette printers. Extra large textured writing surface Wide finger tab on the back of the cassette for easy opening The lid stays attached for repeated opening and closing
This item is not returnable. View return policy
Catalog No. 50-146-0344 Supplier Thomas Scientific Supplier No. 1202D82
The 35Ý angled cassette features a large writing surface and two side panels ideal for labeling. The one-piece design allows for the lids to be opened and closed repeatedly without the lid getting detached from the base. The lid securely locks into the base and keeps samples safe and secure. The 35Ý angled cassettes can be manually loaded into most cassette printers. Extra large textured writing surface Wide finger tab on the back of the cassette for easy opening The lid stays attached for repeated opening and closing