Learn More
Heidolph™ Cassette Without Flask for Hei-VAP Industrial Rotary Evaporator
SDP
Catalog No. 13900952 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Hei-VAP Industrial Rotary Evaporator
Catalog No. For Use With (Equipment)
13-900-952 Hei-VAP Industrial Rotary Evaporator
1 options
Catalog No. 13-900-952 Supplier Heidolph™ Supplier No. 036280116
Add to Cart
Add to Cart
Cassette without flask guards against glassware breakage while providing a self-standing support for receiving flask.
Included with Hei-VAP Industrial Rotary Evaporator
Specifications
Type Enclosure
For Use With (Equipment) Hei-VAP Industrial Rotary Evaporator
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov