Learn More
Enterprise Technology Solutions Cassida Bill Counter,Back Load,UV Sensor,9-3/4"x11-1/2"x7-1/2",GY
SDP
Catalog No. 5000833227
Encompass_Preferred
5520 UV/MG Currency Counter is a professional currency counter designed to save time and simplify operation to take on a variety of cash-processing jobs. It offers intuitive operation, simple controls and automatic features that are easy to operate. Infrared sensors are able to spot a wide range of bill discrepancies including, half notes, double bills and chain coins. It will stop the count, alert you with a beep and display the error type of the screen. UV sensors are able to detect a wide range of counterfeit bills. Currency counter stops and dispenses the bill into its stacker for removal. Professional-grade design counts 1,300 bills per minute. Lightweight, compact construction with built-in handle is easy to transport. Contemporary style blends with any business environment.
Catalog No. 50-008-33227 Supplier Enterprise Technology Solutions Supplier No. CDA5520UVMG
5520 UV/MG Currency Counter is a professional currency counter designed to save time and simplify operation to take on a variety of cash-processing jobs. It offers intuitive operation, simple controls and automatic features that are easy to operate. Infrared sensors are able to spot a wide range of bill discrepancies including, half notes, double bills and chain coins. It will stop the count, alert you with a beep and display the error type of the screen. UV sensors are able to detect a wide range of counterfeit bills. Currency counter stops and dispenses the bill into its stacker for removal. Professional-grade design counts 1,300 bills per minute. Lightweight, compact construction with built-in handle is easy to transport. Contemporary style blends with any business environment.