Learn More
Electron Microscopy Sciences IP Biopsy VI Jet Biopsy VI Cassettes, Gold
SDP
Catalog No. 501926953
Encompass_Preferred
IP Biopsy VI cassettes are two-piece biopsy cassettes with 0.67 mm pores, a 45 degree writing surface and a secure latch closure. Taped cassettes have been designed for the Leica IPC Cassette Printer and offer <1% printing error rate. Loose: CE approved weight-5 Kg, dimensions inches (L x W x H)-12 x 7 x 10. S
Color: Gold
This item is not returnable. View return policy
Catalog No. 50-192-6953 Supplier Electron Microscopy Sciences Supplier No. 70076124
IP Biopsy VI cassettes are two-piece biopsy cassettes with 0.67 mm pores, a 45 degree writing surface and a secure latch closure. Taped cassettes have been designed for the Leica IPC Cassette Printer and offer <1% printing error rate. Loose: CE approved weight-5 Kg, dimensions inches (L x W x H)-12 x 7 x 10. S
Color: Gold