Learn More
Electron Microscopy Sciences Supa Mega Mother Ship Cassette, Red
SDP
Catalog No. 501928702
Encompass
The Supa Mega Mother Ship Cassette allows the use of a standard tissue processing cassette to clearly print a 2D barcode and test information, eliminating risks associated with hand writing on cassettes. Designed for using a docking area for standard processing of cassettes.
Improved TAT, significantly reducing processing times
Improved specimen quality, reducing distortion with no curling or "pringle" effects
Time saving by ensuring rapid casting of block, 30 mins at -8 deg C using a Supa Mega Mother Ship base mold
Improved processing with a large open area hex pore design to maximize fluid exchange
Reduced carry-over during tissue processing
Enhanced sectioning quality through greater adhesion of the paraffin block to the cassette
Elimination of the possibility of the embedded specimen being separated from the cassette when sectioning fibrous specimens
Color: Red
Catalog No. 50-192-8702 Supplier Electron Microscopy Sciences Supplier No. 62511R
The Supa Mega Mother Ship Cassette allows the use of a standard tissue processing cassette to clearly print a 2D barcode and test information, eliminating risks associated with hand writing on cassettes. Designed for using a docking area for standard processing of cassettes.
Improved TAT, significantly reducing processing times
Improved specimen quality, reducing distortion with no curling or "pringle" effects
Time saving by ensuring rapid casting of block, 30 mins at -8 deg C using a Supa Mega Mother Ship base mold
Improved processing with a large open area hex pore design to maximize fluid exchange
Reduced carry-over during tissue processing
Enhanced sectioning quality through greater adhesion of the paraffin block to the cassette
Elimination of the possibility of the embedded specimen being separated from the cassette when sectioning fibrous specimens
Color: Red