Learn More
Med Vet International Cast Cutter, Accessory: Sagittal Blade 12x7x1.2mm, J0866B
Catalog No. 5000584774
Encompass_Preferred
Designed with multiple attachments that allow for a wide range of orthopedic procedures, as well as cast removal. The oscillating head permits delicate work or more aggressive cutting as needed.Suggested procedures:Femoral head excisionTriple Pelvic Osteotomy TPOAmputationsTrochlear wedge osteotomyNecropsyCast removalTibial wedge osteotomySaw specifications:Oscillations: 14,500/min.9.6V, includes 110V charger and battery
Catalog No. 50-005-84774 Supplier Med Vet International Supplier No. JOR866B
Designed with multiple attachments that allow for a wide range of orthopedic procedures, as well as cast removal. The oscillating head permits delicate work or more aggressive cutting as needed.Suggested procedures:Femoral head excisionTriple Pelvic Osteotomy TPOAmputationsTrochlear wedge osteotomyNecropsyCast removalTibial wedge osteotomySaw specifications:Oscillations: 14,500/min.9.6V, includes 110V charger and battery