Learn More
Med Vet International Cast Cutter blades, Accessory: Saw blades 0.8mm thick 19.5mm, J0866F
Catalog No. 5000584696
Encompass_Preferred
0.8mm cut thickness, 19.5mm width, 41mm length. Great blades for TTA or TTO procedure
Catalog No. 50-005-84696 Supplier Med Vet International Supplier No. JOR866F
0.8mm cut thickness, 19.5mm width, 41mm length. Great blades for TTA or TTO procedure