Learn More
Med Vet International Bsn Specialist Cast Padding 2" X 4Yd, 24/Pack
Catalog No. 502832224
Encompass_Preferred
Cotton and Rayon blend for excellent absorption. Micropleated design results in conformably, protection and ease of application. Specialist cast padding provides exceptional under-cast protection Excellent conformability. Contours smoothly over prominences to provide a more comfortable healing process. Padding maximizes the wicking of body moisture under a cast. Unique cotton blend construction reduces tucking and tearing. Rayon provides high-tensile strength while cotton enables superior absorbency. BSN Specialist Cast Padding also available in 3"x4" YDS (9043), 4"x4" YDS (9044), 6"x4" YDS (9046).
Catalog No. 50-283-2224 Supplier Med Vet International Supplier No. 9062
Cotton and Rayon blend for excellent absorption. Micropleated design results in conformably, protection and ease of application. Specialist cast padding provides exceptional under-cast protection Excellent conformability. Contours smoothly over prominences to provide a more comfortable healing process. Padding maximizes the wicking of body moisture under a cast. Unique cotton blend construction reduces tucking and tearing. Rayon provides high-tensile strength while cotton enables superior absorbency. BSN Specialist Cast Padding also available in 3"x4" YDS (9043), 4"x4" YDS (9044), 6"x4" YDS (9046).