Learn More
Med Vet International Specialist Cast Padding, 3"X4Yds, 12/Pack
Catalog No. 502830938
Encompass_Preferred
Made from 100% cotton fibers, Specialist 100 cast padding is designed to provide under-cast protection and wick away body exudates while offering the comfort and breathability of pure cotton. This cast padding fits body contours without slipping, maximizing patient protection and comfort throughout the healing process.
Catalog No. 50-283-0938 Supplier Med Vet International Supplier No. 9083
Made from 100% cotton fibers, Specialist 100 cast padding is designed to provide under-cast protection and wick away body exudates while offering the comfort and breathability of pure cotton. This cast padding fits body contours without slipping, maximizing patient protection and comfort throughout the healing process.