Learn More
Enzo Life Sciences Castanospermine (100 mg). CAS: 79831-76-8
SDP
Supplier:  Enzo Life Sciences BMLS1070100
Encompass_Preferred
Plant alkaloid. Potent inhibitor of α- and β-glucosidases. Exhibits antiviral properties. Inhibits thyroglobulin secretion. Antitumor activity. Inhibits protein glycosylation. Prevents angiogenesis. Purity: ≥95% (TLC). Solubility: Soluble in water. Long Term Storage: +4°C.
Catalog No. 50-201-1733