Learn More
MSC Caster Connection Rigid Top Plate Caster, Polyurethane, 5 in Wheel, 450 lb Capacity
Catalog No. 5000527825
Encompass
This rigid top plate caster from Caster Connection features a durable polyurethane wheel with a 5 inch diameter and a load capacity of 450 lb. It is designed for applications requiring optimal maneuverability and offers quiet operation and excellent ergonomic performance.
  • Extremely quiet operation
  • Rejects floor debris
  • Reduces downtime and maintenance costs
  • Excellent ergonomic performance
  • Low push/pull forces required for movement
Catalog No. 50-005-27825 Supplier MSC Supplier No. 49080906
This rigid top plate caster from Caster Connection features a durable polyurethane wheel with a 5 inch diameter and a load capacity of 450 lb. It is designed for applications requiring optimal maneuverability and offers quiet operation and excellent ergonomic performance.
  • Extremely quiet operation
  • Rejects floor debris
  • Reduces downtime and maintenance costs
  • Excellent ergonomic performance
  • Low push/pull forces required for movement