Learn More
MSC Caster Connection Swivel Top Plate Caster, Polyurethane, 4 in Wheel
Catalog No. 5000512576
Encompass
This swivel top plate caster features a polyurethane wheel with a 4 in diameter and 2 in width. It has a 700 lb capacity and an overall height of 5-5/8 in. The caster also includes a side brake and a 4.5 x 4 in plate.
  • Side brake for secure positioning.
  • Plate mount type for easy installation.
  • Equipped with sealed precision ball bearings.
  • Kingpinless swivel construction for enhanced durability.
  • Zinc finish provides corrosion resistance.
  • Extremely quiet operation with an extended swivel lead.
  • Reduces downtime and maintenance costs.
  • Offers excellent ergonomic performance and requires low push/pull forces for movement.
  • Designed for optimal maneuverability in applications requiring frequent movement.
Catalog No. 50-005-12576 Supplier MSC Supplier No. 49080732
This swivel top plate caster features a polyurethane wheel with a 4 in diameter and 2 in width. It has a 700 lb capacity and an overall height of 5-5/8 in. The caster also includes a side brake and a 4.5 x 4 in plate.
  • Side brake for secure positioning.
  • Plate mount type for easy installation.
  • Equipped with sealed precision ball bearings.
  • Kingpinless swivel construction for enhanced durability.
  • Zinc finish provides corrosion resistance.
  • Extremely quiet operation with an extended swivel lead.
  • Reduces downtime and maintenance costs.
  • Offers excellent ergonomic performance and requires low push/pull forces for movement.
  • Designed for optimal maneuverability in applications requiring frequent movement.