Learn More
MSC Caster Connection Swivel Top Plate Caster, Polyurethane on Aluminum, 5 in Wheel Dia, 750 lb Capacity
Catalog No. 5000535114
Encompass
This swivel top plate caster features a polyurethane on aluminum wheel, designed for robust performance. It offers a 750 lb load capacity and kingpinless swivel construction for smooth movement.
  • 5 in wheel diameter
  • 2 in wheel width
  • 750 lb load capacity
  • 6-1/2 in overall height
  • 4.5 in plate length
  • 4 in plate width
  • Polyurethane on aluminum wheel material
  • Sealed precision ball bearing
  • Kingpinless swivel construction
Catalog No. 50-005-35114 Supplier MSC Supplier No. 47173042
This swivel top plate caster features a polyurethane on aluminum wheel, designed for robust performance. It offers a 750 lb load capacity and kingpinless swivel construction for smooth movement.
  • 5 in wheel diameter
  • 2 in wheel width
  • 750 lb load capacity
  • 6-1/2 in overall height
  • 4.5 in plate length
  • 4 in plate width
  • Polyurethane on aluminum wheel material
  • Sealed precision ball bearing
  • Kingpinless swivel construction