Learn More
MSC Caster Connection Swivel Top Plate Caster, Polyurethane on Aluminum, 6 in Wheel, 1000 lb
Catalog No. 5000533035
Encompass
This swivel top plate caster is designed for demanding applications, featuring a durable polyurethane on aluminum wheel. With a 6 inch wheel diameter and a robust 1000 lb load capacity, it ensures reliable performance and smooth mobility. The kingpinless swivel construction provides enhanced maneuverability and stability.Key features include:
  • Polyurethane on aluminum wheel material.
  • 6 inch wheel diameter.
  • 2 inch wheel width.
  • 1000 lb load capacity.
  • 7-1/2 inch overall height.
  • Plate mount type.
  • Sealed precision ball bearing type.
  • Kingpinless swivel construction.
Catalog No. 50-005-33035 Supplier MSC Supplier No. 47172945
This swivel top plate caster is designed for demanding applications, featuring a durable polyurethane on aluminum wheel. With a 6 inch wheel diameter and a robust 1000 lb load capacity, it ensures reliable performance and smooth mobility. The kingpinless swivel construction provides enhanced maneuverability and stability.Key features include:
  • Polyurethane on aluminum wheel material.
  • 6 inch wheel diameter.
  • 2 inch wheel width.
  • 1000 lb load capacity.
  • 7-1/2 inch overall height.
  • Plate mount type.
  • Sealed precision ball bearing type.
  • Kingpinless swivel construction.