Learn More
Grainger GRAINGER APPROVED Rigid Plate Caster
Supplier:  Grainger 45XC26
Encompass_Preferred
The Grainger Approved Rigid Plate Caster is a 4" diameter rigid plate caster with a zinc plated steel top plate and a gray thermoplastic rubber wheel. It has a load capacity of 250 lbs per caster and features a sealed precision ball bearing axle for smooth rolling. This rigid caster is ideal for material handling equipment, workbenches, carts, and other applications requiring a heavy duty caster. The top mounting plate has holes to facilitate bolting or welding it to the equipment frame. Overall this is a heavy duty rigid plate caster well suited for industrial applications.
Catalog No. NC2815442