Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ Casters for 6 Gallon Buckets

Catalog No. 19200710 Shop All Vileda Professional Products
Catalog No. 19-200-710 Supplier Vileda Professional™ Supplier No. 135083
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Casters are for use with 6 gallon buckets

  • Not autoclavable
  • For use with #131586 and #131584
  • Sold separately from buckets

Specifications

Autoclavable Not Autoclavable
Color Gray
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.