Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Casting Material Mixing Bags
SDP

Catalog No. 22446189 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W):
13 x 18 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-446-189 13 x 18 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-446-189 Supplier Mopec Supplier No. FS123
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for preparing and handling casting materials in the field or lab

  • Features a resealable design with a thickness of 4 mm.
  • Provides a clean and simple way to mix water with casting material.
  • Made from tough, flexible plastic for reliable handling.
  • Allows easy mixing, pouring, and storage.
  • Helps minimize mess and material loss.
  • Ideal for forensic casting applications.
  • Suitable for footprint and tire impression evidence.

Specifications

Dimensions (L x W) 13 x 18 in.
Product Type Evidence Bag
Color Transparent
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.