Description
- Kit includes a mixing card, spatula, catalyst, casting material, and casting putty
- Designed for highly detailed casting of tool and jimmy marks
- Renders small details clearly
- Comes in various colors, mixes quickly, sets in minutes
- Provides high contrast for microscopic observations
- Performs in Below-freezing temperatures
- Putty and catalyst are mixed on a card with a spatula before application
- Casts typically set within 6 to 8 minutes
- In Below-freezing temperatures, casts typically set within 15 minutes
- Black casting putty is used with white, silver, or fluorescent powders
- White casting putty is used with black powders
- Gray casting putty provides contrast on Light-colored surfaces and with side-lighting
- Brown casting putty helps increase visibility of cast details
- Compatible accessory: FS396 Additional Tube of Catalyst
Specifications
Specifications
| Product Type | Casting Putty |
| Color | White |