Learn More
Cayman Chemical Castanospermine, Purity: ≥98%, CAS: 79831-76-8, 10 mg
SDP
Catalog No. 502953269
Encompass_Preferred
An inhibitor of both α- and β-glucosidases, inhibiting lysosomal and neutral α-glucosidases with Ki values of 0.1 and 10 μM, respectively, and lysosomal and cytosolic β-glucosidases with Ki values of 7 and 40 μM, respectively; blocks N-linked glycosylation, affecting protein trafficking
Catalog No. 50-295-3269 Supplier Cayman Chemical Supplier No. 1131310 MG
An inhibitor of both α- and β-glucosidases, inhibiting lysosomal and neutral α-glucosidases with Ki values of 0.1 and 10 μM, respectively, and lysosomal and cytosolic β-glucosidases with Ki values of 7 and 40 μM, respectively; blocks N-linked glycosylation, affecting protein trafficking