Learn More
Selleck Chemical LLC Castor oil
SDP
Catalog No. 5000002895
Encompass_Preferred
Castor oil is a triglyceride derived from Ricinus communis and has been frequently used in antidiarrhea bioassays on laboratory animals It has been used to study the effects on rat lens conditions Castor oil has also been used as a test reagent in the rat model of castor oil-induced diarrhoea to study the dose-response relationship and mechanism of action of Saccharomyces boulardii
Catalog No. 50-000-02895 Supplier Selleck Chemical LLC Supplier No. S6700100ML
Castor oil is a triglyceride derived from Ricinus communis and has been frequently used in antidiarrhea bioassays on laboratory animals It has been used to study the effects on rat lens conditions Castor oil has also been used as a test reagent in the rat model of castor oil-induced diarrhoea to study the dose-response relationship and mechanism of action of Saccharomyces boulardii