Learn More
Medchemexpress LLC Castor oil | 8001-79-4 | MFCD00130746 | Mixture of triglycerides (no single formula) | 100 ML
SDP
Catalog No. 5000241666
Encompass_Preferred
Castor oil is a natural triglyceride derived from Ricinus communis seeds, supplied for laboratory research as a multifunctional solvent and raw material. It is used as a solvent, co-solvent, stabilizing agent, and polyol precursor for making polymer-nanoparticle composites, and finds application in cosmetic formulation research, lubricant and biofuel studies, and material coatings. For research use only; not for human or clinical use.
  • Natural triglyceride and vegetable oil.
  • Used as a solvent and co-solvent in laboratory applications.
  • Functions as a stabilizing agent and polyol precursor for polymers.
  • Applicable to cosmetic, lubricant, biofuel, and coating research.
  • Appearance: oil, colorless to light yellow.
  • Available in small pack sizes suitable for lab use.
Catalog No. 50-002-41666 Supplier Medchemexpress LLC Supplier No. HY107799100ML
Add to Cart
Add to Cart
Castor oil is a natural triglyceride derived from Ricinus communis seeds, supplied for laboratory research as a multifunctional solvent and raw material. It is used as a solvent, co-solvent, stabilizing agent, and polyol precursor for making polymer-nanoparticle composites, and finds application in cosmetic formulation research, lubricant and biofuel studies, and material coatings. For research use only; not for human or clinical use.
  • Natural triglyceride and vegetable oil.
  • Used as a solvent and co-solvent in laboratory applications.
  • Functions as a stabilizing agent and polyol precursor for polymers.
  • Applicable to cosmetic, lubricant, biofuel, and coating research.
  • Appearance: oil, colorless to light yellow.
  • Available in small pack sizes suitable for lab use.