Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Castor Oil, Sulfated, Spectrum™ Chemical
SDP

Catalog No. 18601506 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-506 4 L Amber Glass Bottle
18-601-507 20 L Amber Glass Bottle
18-601-508 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-601-506 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CA2094LTGL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

CA209, 8002-33-3

Spectrum™ Chemical Castor Oil, Sulfated is an oil that is derived from the castor beans and is made by adding sulfuric acid to the oil. A colorless oil that has no taste or smell and boils at 313°C, it is used as a synthetic detergent in formulating lubricants, softeners, and dyeing assistants. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 8002-33-3 , 7732-18-5

Specifications

CAS Max % 68-76%, 24-32%
Packaging Amber Glass Bottle
Quantity 4 L
Grade Reagent
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Castor Oil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.