Learn More
Med Vet International Castration Bands, Latex, 100 Count
Catalog No. 502828815
Encompass_Preferred
Castrating bands/rings are applied with a castrating band applicator for tail docking or castration. Made of premium Malaysian latex. Excellent pressure ratings and retention levels. Store in a cool, dark place.
Catalog No. 50-282-8815 Supplier Med Vet International Supplier No. PHVCASTBAND100
Castrating bands/rings are applied with a castrating band applicator for tail docking or castration. Made of premium Malaysian latex. Excellent pressure ratings and retention levels. Store in a cool, dark place.