Learn More
Electron Microscopy Sciences Castro Viejo Needle Holder Model PCF FX
SDP
Catalog No. 5030632
Encompass_Preferred
5.5" long, straight tips: 1.03mm wide x 4mm long.
Model: PCF FX
Catalog No. 50-306-32 Supplier Electron Microscopy Sciences Supplier No. 77890FX
5.5" long, straight tips: 1.03mm wide x 4mm long.
Model: PCF FX