Learn More
Electron Microscopy Sciences Castro Viejo Needle Holder Model PCF-TCL
SDP
Catalog No. 5030633
Encompass_Preferred
Straight 5mm serrated jaws, comes with a mechanism lock.
Model: PCF-TCL
Catalog No. 50-306-33 Supplier Electron Microscopy Sciences Supplier No. 77890S
Straight 5mm serrated jaws, comes with a mechanism lock.
Model: PCF-TCL