Learn More
Roboz Surgical CASTROVIEJO 3 1/2IN (9 CM)
SDP
Catalog No. NC2132011
Encompass
Castroviejo 3 1/2” (9 cm) Long 9X0.15mm (Cutting Edge Length X Tip Width) Straight Sharp SureCut
This item is not returnable. View return policy
Catalog No. NC2132011 Supplier Roboz Surgical Supplier No. RS5650SC
Castroviejo 3 1/2” (9 cm) Long 9X0.15mm (Cutting Edge Length X Tip Width) Straight Sharp SureCut