Learn More
Roboz Surgical CASTROVIEJO 9X.3MM ANG S/S
SDP
Catalog No. NC9192103
Encompass
Castroviejo Micro Dissecting Spring Scissors; Angular (Points To Left), Sharp Points; 9mm Cutting Edge; 0.15mm Tip Width; 3-1/2 in. Overall Length Angled blades make it easier to cut from above while keeping the blades aligned with the line of cutting
Catalog No. NC9192103 Supplier Roboz Surgical Supplier No. RS5658
Castroviejo Micro Dissecting Spring Scissors; Angular (Points To Left), Sharp Points; 9mm Cutting Edge; 0.15mm Tip Width; 3-1/2 in. Overall Length Angled blades make it easier to cut from above while keeping the blades aligned with the line of cutting