Learn More
Roboz Surgical CASTROVIEJO 9X.3MM STR S/S
SDP
Catalog No. NC9457429
Encompass
Castroviejo Micro Dissecting Spring Scissors; Straight, Sharp Points; 9mm Cutting Edge; 0.15mm Tip Width; 3-1/2 in. Overall Length
Catalog No. NC9457429 Supplier Roboz Surgical Supplier No. RS5650
Castroviejo Micro Dissecting Spring Scissors; Straight, Sharp Points; 9mm Cutting Edge; 0.15mm Tip Width; 3-1/2 in. Overall Length