Learn More
World Precision Instrument Castroviejo Curved Scissors, 12cm, Blunt, 11mm Tips, Titanium
SDP
Supplier:  World Precision Instrument WP2120R
Encompass
Castroviejo titanium scissors are surgical scissors designed for precise cutting in delicate surgical procedures like ophthalmic surgery, microsurgery, microdissection, and other procedures requiring precision cutting. Titanium Castroviejo scissors are 40% lighter than the stainless-steel varieties, reducing hand fatigue. Titanium is known for its strength, corrosion resistance, and biocompatibility. It is non-magnetic, and it won’t rust.  Castroviejo scissors are spring scissors with a fine and slender design, allowing for precise and controlled cutting. The blades are sharp and tapered, making them suitable for intricate tasks. These two sizes of Castroviejo titanium scissors have curved blades. One has 16 mm tips and the other has 11 mm blunted tips. Curved blades are often used for tasks requiring a more angled approach. The tips of Castroviejo scissors are fine, allowing for precise entry into tissues without causing unnecessary trauma.
Catalog No. 50-253-8330