Learn More
Fine Science Tools Castroviejo Micro Needle Holders, Curved, 9 cm
SDP
Catalog No. NC0126754
Encompass
Product: Castroviejo Micro Needle Holder - Curved/Smooth/9cm/with Lock/ Tip Shape: Curved/ Tip Width: 0.6mm/ Clamping Length: 5mm/ Scissors: No/ Lock: Yes/ Alloy: Stainless Steel/ Length: 9cm/ Serrated: No/ Unit: Each
Catalog No. NC0126754 Supplier Fine Science Tools Supplier No. 1206101
Product: Castroviejo Micro Needle Holder - Curved/Smooth/9cm/with Lock/ Tip Shape: Curved/ Tip Width: 0.6mm/ Clamping Length: 5mm/ Scissors: No/ Lock: Yes/ Alloy: Stainless Steel/ Length: 9cm/ Serrated: No/ Unit: Each