Learn More
Fine Science Tools Castroviejo Micro Needle Holders, Straight, 9 cm
SDP
Catalog No. NC0655805
Encompass
Product: Castroviejo Micro Needle Holder/9cm/ Tip Shape: Straight/ Tip Width: 0.6mm/ Clamping Length: 5mm/ Scissors: No/ Lock: No/ Alloy: Stainless Steel/ Length: 9cm/ Serrated: No/ Unit: Each
Catalog No. NC0655805 Supplier Fine Science Tools Supplier No. 1206002
Product: Castroviejo Micro Needle Holder/9cm/ Tip Shape: Straight/ Tip Width: 0.6mm/ Clamping Length: 5mm/ Scissors: No/ Lock: No/ Alloy: Stainless Steel/ Length: 9cm/ Serrated: No/ Unit: Each