Learn More
Stoelting™ Castroviejo Retractor
SDP
Catalog No. 10000490 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Retractor, Scew-type, Adjustable
Catalog No. Model
10-000-490 Retractor, Scew-type, Adjustable
1 options
Catalog No. 10-000-490 Supplier Stoelting™ Supplier No. 52127P
Add to Cart
Add to Cart
Stoelting™ Castroviejo Retractor can be used to actively separate the edges of a surgical incision or wound, and can hold back underlying organs and tissues, so that body parts under the incision may be accessed.
  • Sharp retractors are better at providing a secure grip and they can safely hold back the skin, muscles, fascia, tendons, or the periosteum but they may damage vessels, nerves, the peritoneum, glands or cysts.
  • Blunt retractors are recommended when iatrogenic injury is a concern.
  • The main benefit of blunt retractors is that they prevent unintended injury to the tissue held by the retractor or to the underlying tissue.
Specifications
For Use With (Application) Neuroscience
Length (English) 3.75 in.
Model Retractor, Scew-type, Adjustable