Learn More
Fine Science Tools Castroviejo Spring Scissors - Sharply Curved Up
SDP
Catalog No. NC9300426
Encompass
Tip Shape: Curved/ Tips: Sharp/ Cutting Edge: 10mm/ Tip Diameter: 0.255mm/ Alloy / Material: Stainless Steel/ Length: 10cm/ Serrated: No
Catalog No. NC9300426 Supplier Fine Science Tools Supplier No. 1501710
Tip Shape: Curved/ Tips: Sharp/ Cutting Edge: 10mm/ Tip Diameter: 0.255mm/ Alloy / Material: Stainless Steel/ Length: 10cm/ Serrated: No