Learn More
Fine Science Tools Castroviejo Micro Needle Holders, Straight, 9 cm
SDP
Catalog No. NC9162912
Encompass
Product: Castroviejo Micro Needle Holder - Straight/Smooth/9cm/with Lock/ Tip Shape: Straight/ Tip Width: 0.6mm/ Clamping Length: 5mm/ Scissors: No/ Lock: Yes/ Alloy: Stainless Steel/ Length: 9cm/ Serrated: Yes/ Unit: Each
Catalog No. NC9162912 Supplier Fine Science Tools Supplier No. 1206001
Product: Castroviejo Micro Needle Holder - Straight/Smooth/9cm/with Lock/ Tip Shape: Straight/ Tip Width: 0.6mm/ Clamping Length: 5mm/ Scissors: No/ Lock: Yes/ Alloy: Stainless Steel/ Length: 9cm/ Serrated: Yes/ Unit: Each