Learn More
Fine Science Tools Castroviejo Surgical Caliper
SDP
Catalog No. NC1750377
Encompass
Castroviejo Surgical Caliper, Increments: 0.5mm, Maximum Measurement: 20mm, Alloy / Material: Stainless Steel, Length: 8.5cm, Used for precise measurement during surgery
This item is not returnable. View return policy
Catalog No. NC1750377 Supplier Fine Science Tools Supplier No. 1800035
Castroviejo Surgical Caliper, Increments: 0.5mm, Maximum Measurement: 20mm, Alloy / Material: Stainless Steel, Length: 8.5cm, Used for precise measurement during surgery