Learn More
World Precision Instrument Castroviejo Suturing Forceps, 11 cm, Straight
SDP
Catalog No. 502538301
Encompass
  • Straight tips, 5.5mm, Tying platform
  • Wide serrated handle
Catalog No. 50-253-8301 Supplier World Precision Instrument Supplier No. WP5025501
  • Straight tips, 5.5mm, Tying platform
  • Wide serrated handle