Learn More
Fine Science Tools Castroviejo Needle Holder, Straight, 14 cm
SDP
Catalog No. NC9147991
Encompass
Product: Castroviejo Needle Holder/ Tip Shape: Straight/ Tip Width: 1.5mm/ Clamping Length: 10mm/ Scissors: No/ Lock: Yes/ Alloy: Stainless Steel/ Length: 14cm/ Serrated: Yes/ Feature: Tungsten Carbide/ Unit: Each
Catalog No. NC9147991 Supplier Fine Science Tools Supplier No. 1256514
Product: Castroviejo Needle Holder/ Tip Shape: Straight/ Tip Width: 1.5mm/ Clamping Length: 10mm/ Scissors: No/ Lock: Yes/ Alloy: Stainless Steel/ Length: 14cm/ Serrated: Yes/ Feature: Tungsten Carbide/ Unit: Each